missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-acetyl-α-D-galactosamine, 99%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210006405
Specifications
N-Acetyl-α-D-galactosamine | |
White | |
C8H15NO6 | |
n-acetyl-d-galactosamine, galnac, 2-acetamido-2-deoxy-d-galactopyranose, d-galnac, 2-acetamido-2-deoxy-d-galactose, n-acetyl-d-chondrosamine, n-3r,4r,5r,6r-2,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-3-yl acetamide, n-acetylgalactosamine, acetylgalactosamine, n-acetyl-galactosamine | |
CC(=O)NC1C(C(C(OC1O)CO)O)O | |
221.209 | |
CHEBI:28497 | |
Crystalline Powder |
14215-68-0 | |
0.99 | |
5 g | |
OVRNDRQMDRJTHS-JAJWTYFOSA-N | |
N-[(2R,3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide | |
440552 | |
221.2 |
Chemical Identifiers
14215-68-0 | |
221.209 | |
n-acetyl-d-galactosamine, galnac, 2-acetamido-2-deoxy-d-galactopyranose, d-galnac, 2-acetamido-2-deoxy-d-galactose, n-acetyl-d-chondrosamine, n-3r,4r,5r,6r-2,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-3-yl acetamide, n-acetylgalactosamine, acetylgalactosamine, n-acetyl-galactosamine | |
CHEBI:28497 | |
CC(=O)NC1C(C(C(OC1O)CO)O)O |
C8H15NO6 | |
OVRNDRQMDRJTHS-JAJWTYFOSA-N | |
440552 | |
N-[(2R,3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |