missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-acetyl-D-glucosamine, 100%, MP Biomedicals™
$70.56 - $272.79
Chemical Identifiers
CAS | 7512-17-6 |
---|---|
Molecular Formula | C8H15NO6 |
Molecular Weight (g/mol) | 221.21 |
MDL Number | MFCD00061615 |
InChI Key | OVRNDRQMDRJTHS-WZWLWIOANA-N |
Synonym | 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc |
IUPAC Name | N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |
SMILES | CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10006825
|
MP Biomedicals Inc
0210006825 |
25 g |
Each for $70.56
|
|
|||||
ICN10006880
|
MP Biomedicals Inc
0210006880 |
100 g |
Each for $272.79
|
|
|||||
Chemical Identifiers
7512-17-6 | |
221.21 | |
OVRNDRQMDRJTHS-WZWLWIOANA-N | |
N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |
C8H15NO6 | |
MFCD00061615 | |
2-Acetamido-2-deoxy-D-glucose, D-GlcNAc | |
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O |
Specifications
7512-17-6 | |
1 | |
MFCD00061615 | |
2-Acetamido-2-deoxy-D-glucose, D-GlcNAc | |
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O | |
221.21 | |
Fine Crystalline Powder |
White | |
C8H15NO6 | |
25 g | |
OVRNDRQMDRJTHS-WZWLWIOANA-N | |
N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide | |
221.2 | |
N-Acetyl-D-glucosamine |