missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals N-Acetyl-DL-methionine, 99%

Supplier:  Thermo Scientific Chemicals B2186630

 View more versions of this product

Catalog No. AAB2186630


Explore available promotions

Description

Description

N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.

Solubility
Soluble in water, ethanol, ethyl acetate.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications

Specifications

113°C to 117°C
1115-47-5
MFCD00008681
14,96
Soluble in water,ethanol,ethyl acetate.
CSCCC(NC(C)=O)C(O)=O
250 g
6180
99%
N-Acetyl-DL-methionine
C7H13NO3S
1725554
n-acetyl-dl-methionine, ac-dl-met-oh, n-acetylmethionine, acetyl-dl-methionine, dl-n-acetylmethionine, methionamine, methionine, n-acetyl, dl-methionine, n-acetyl, methionine, n-acetyl-, dl, dl-acetylmethionine
XUYPXLNMDZIRQH-UHFFFAOYNA-N
2-acetamido-4-methylsulfanylbutanoic acid
191.25
191.25

Chemical Identifiers

1115-47-5
191.25
XUYPXLNMDZIRQH-UHFFFAOYNA-N
6180
C7H13NO3S
MFCD00008681
n-acetyl-dl-methionine, ac-dl-met-oh, n-acetylmethionine, acetyl-dl-methionine, dl-n-acetylmethionine, methionamine, methionine, n-acetyl, dl-methionine, n-acetyl, methionine, n-acetyl-, dl, dl-acetylmethionine
CSCCC(NC(C)=O)C(O)=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-224-3

RTECSNumber : PD0500000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only