missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-acetyl-DL-tryptophan, ∽99%, MP Biomedicals™
N-acetyl-DL-tryptophan, is used as stabilizer in the human blood-derived therapeutic products normal serum albumin and plasma protein fraction
Supplier: MP Biomedicals Inc 0210014225
Specifications
N-acetyl-DL-tryptophan | |
White | |
C13H14N2O3 | |
n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl | |
CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O | |
25 g | |
2002 | |
246.3 |
87-32-1 | |
∽99% | |
MFCD00005644 | |
DZTHIGRZJZPRDV-UHFFFAOYSA-N | |
2-acetamido-3-(1H-indol-3-yl)propanoic acid | |
246.27 | |
CHEBI:70976 | |
Powder |
Chemical Identifiers
87-32-1 | |
246.27 | |
DZTHIGRZJZPRDV-UHFFFAOYSA-N | |
2002 | |
2-acetamido-3-(1H-indol-3-yl)propanoic acid |
C13H14N2O3 | |
MFCD00005644 | |
n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl | |
CHEBI:70976 | |
CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O |
Safety and Handling
EINECSNumber : 201-739-3
Recommended Storage : Store at +4°C, Store Desiccated.