missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-acetyl-L-cysteine, >96%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210009891
Specifications
N-acetyl-L-cysteine | |
White | |
C5H9NO3S | |
n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit | |
CC(=O)NC(CS)C(=O)O | |
(2R)-2-acetamido-3-sulfanylpropanoic acid | |
163.191 | |
CHEBI:28939 | |
Crystalline Powder |
616-91-1 | |
>96% | |
MFCD00004880 | |
PWKSKIMOESPYIA-BYPYZUCNSA-N | |
1.295g/cm3 (L) | |
1 kg | |
12035 | |
163.2 |
Chemical Identifiers
616-91-1 | |
163.191 | |
PWKSKIMOESPYIA-BYPYZUCNSA-N | |
12035 | |
(2R)-2-acetamido-3-sulfanylpropanoic acid |
C5H9NO3S | |
MFCD00004880 | |
n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit | |
CHEBI:28939 | |
CC(=O)NC(CS)C(=O)O |