missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals N-Acetyl-L-cysteine, 98%

Supplier:  Thermo Scientific Chemicals 160280250

 View more versions of this product

Catalog No. AC160280250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Acetyl-L-cysteine
97.5
109.0°C to 111.0°C
0.5% max.
Authentic
Glass bottle
HSCH2CH(NHCOCH3)CO2H
25 g
+ 5.50
Solubility in water: soluble in water.
CC(=O)NC(CS)C(=O)O
163.19
CHEBI:28939
98%
616-91-1
100.0
White
0.5% max.
98%
C5H9NO3S
MFCD00004880
15, 83
n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit
PWKSKIMOESPYIA-BYPYZUCNSA-N
(2R)-2-acetamido-3-sulfanylpropanoic acid
12035
163.19
Crystalline Powder or Crystals

Chemical Identifiers

616-91-1
163.19
PWKSKIMOESPYIA-BYPYZUCNSA-N
12035
(2R)-2-acetamido-3-sulfanylpropanoic acid
C5H9NO3S
MFCD00004880
n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit
CHEBI:28939
CC(=O)NC(CS)C(=O)O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-498-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only