missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-L-glutamine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 451640050
Specifications
| N-Acetyl-L-glutamine | |
| 206°C | |
| 97% | |
| C7H12N2O4 | |
| 5 g | |
| Solubility in water: soluble | |
| CC(=O)N[C@@H](CCC(N)=O)C(O)=O | |
| 188.18 | |
| CHEBI:21553 | |
| 97% |
| 35305-74-9 | |
| 0.2% max | |
| Glass bottle | |
| MFCD00038159 | |
| n-acetyl-l-glutamine, aceglutamide, l-glutamine, n-acetyl, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid, acetylglutamine, n-alpha-acetyl-l-glutamine, n-acetylglutamine, acutil-s, acetyl-l-glutamine | |
| KSMRODHGGIIXDV-YFKPBYRVSA-N | |
| (2S)-4-carbamoyl-2-acetamidobutanoic acid | |
| 182230 | |
| 188.18 |
Chemical Identifiers
| 35305-74-9 | |
| 188.18 | |
| KSMRODHGGIIXDV-YFKPBYRVSA-N | |
| 182230 | |
| CC(=O)N[C@@H](CCC(N)=O)C(O)=O |
| C7H12N2O4 | |
| MFCD00038159 | |
| n-acetyl-l-glutamine, aceglutamide, l-glutamine, n-acetyl, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid, acetylglutamine, n-alpha-acetyl-l-glutamine, n-acetylglutamine, acutil-s, acetyl-l-glutamine | |
| CHEBI:21553 |
RUO – Research Use Only