missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetyl-L-tyrosine ethyl ester monohydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 102750100

 View more versions of this product

Catalog No. AC102750100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Acetyl-L-tyrosine ethyl ester monohydrate
98.5
80°C to 81°C
Authentic
Glass bottle
4-(HO)C6H4CH2CH(NHCOCH3)CO2C2H5·H2O
10 g
n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate
YWAVLHZJMWEYTA-HVPILOLGNA-N
ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate;hydrate
2723594
99%
36546-50-6
100.0
White
98.5% min. (HPLC)
C13H19NO5
MFCD00149093
+ 23
Solubility in water: soluble in cold water. Other solubilities: soluble in hot water,soluble in ethanol
O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O
269.30
269.29
Crystalline Powder

Chemical Identifiers

36546-50-6
269.30
YWAVLHZJMWEYTA-HVPILOLGNA-N
2723594
O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O
C13H19NO5
MFCD00149093
n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate
ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate;hydrate
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only