missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetylcaprolactam, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1825722

 View more versions of this product

Catalog No. AAA1825722


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Acetylcaprolactam
1.094
122°C (252°F)
1.489
100 g
n-acetylcaprolactam, acetylcaprolactam, n-acetyl-6-caprolactam, 2h-azepin-2-one, 1-acetylhexahydro, acetylkaprolaktam, n-acetylhexanelactam, acetylkaprolaktam czech, 1-acetylhexahydro-2h-azepin-2-one, n-acetyl-epsilon-caprolactam, n-acetyl-.epsilon.-caprolactam
CC(=O)N1CCCCCC1=O
155.20
155.2
1888-91-1
124°C (12 mmHg)
C8H13NO2
MFCD00003262
119598
QISSLHPKTCLLDL-UHFFFAOYSA-N
1-acetylazepan-2-one
15904
99%

Chemical Identifiers

1888-91-1
155.20
QISSLHPKTCLLDL-UHFFFAOYSA-N
15904
CC(=O)N1CCCCCC1=O
C8H13NO2
MFCD00003262
n-acetylcaprolactam, acetylcaprolactam, n-acetyl-6-caprolactam, 2h-azepin-2-one, 1-acetylhexahydro, acetylkaprolaktam, n-acetylhexanelactam, acetylkaprolaktam czech, 1-acetylhexahydro-2h-azepin-2-one, n-acetyl-epsilon-caprolactam, n-acetyl-.epsilon.-caprolactam
1-acetylazepan-2-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c

H302-H319-H361

EINECSNumber : 217-565-6

RTECSNumber : CM3599500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only