missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Allylphthalimide, 97%, Thermo Scientific™
$145.32 - $424.92
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH63724MD
|
Thermo Scientific Chemicals
H63724MD |
250 mg |
Each for $145.32
|
|
|||||
AAH6372403
|
Thermo Scientific Chemicals
H6372403 |
1 g |
Each for $424.92
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
5428-09-1 | |
MFCD00158662 |
Specifications
5428-09-1 | |
C11H9NO2 | |
250 mg | |
MHHGQWMCVNQHLG-UHFFFAOYSA-N | |
2-prop-2-enylisoindole-1,3-dione | |
224401 | |
97% | |
N-Allylphthalimide |
White | |
MFCD00158662 | |
2-allylisoindoline-1,3-dione, n-allylphthalimide, n-2-propenyl phthalimide, 2-allyl-1h-isoindole-1,3 2h-dione, 2-prop-2-en-1-yl-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 2-2-propen-1-yl, 2-prop-2-en-1-yl-2,3-dihydro-1h-isoindole-1,3-dione, n-allylphthalimid, n-allyl-phthalimide, 2-allylisoindole-1,3-dione | |
C=CCN1C(=O)C2=CC=CC=C2C1=O | |
187.20 | |
187.19 | |
Solid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures