Learn More
N-Allylthiourea, 98%, Thermo Scientific Chemicals
$69.77 - $155.96
Chemical Identifiers
CAS | 109-57-9 |
---|---|
Molecular Formula | C4H8N2S |
Molecular Weight (g/mol) | 116.182 |
MDL Number | MFCD00004940 |
InChI Key | HTKFORQRBXIQHD-UHFFFAOYSA-N |
Synonym | allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin |
PubChem CID | 1549517 |
ChEBI | CHEBI:74079 |
IUPAC Name | prop-2-enylthiourea |
SMILES | C=CCNC(=S)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0337714
|
Thermo Scientific Chemicals
L0337714 |
25 g |
Each for $69.77
|
|
|||||
AAL0337722
|
Thermo Scientific Chemicals
L0337722 |
100 g |
Each for $155.96
|
|
|||||
Description
N-Allylthiourea is a nitrification inhibitor used in the study on the transformation of diclofenac, naproxen and bisoprolol under aerobic and anaerobic conditions. It is also used in medicine to minimize scar tissue in order to fight against a type of dermatitis. Further, it inhibits the growth of transplanted tumors in mice. It acts as a chelating agent. In addition, it is used in cosmetics, preservative and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Allylthiourea is a nitrification inhibitor used in the study on the transformation of diclofenac, naproxen and bisoprolol under aerobic and anaerobic conditions. It is also used in medicine to minimize scar tissue in order to fight against a type of dermatitis. Further, it inhibits the growth of transplanted tumors in mice. It acts as a chelating agent. In addition, it is used in cosmetics, preservative and in organic synthesis.
Solubility
Soluble in water and alcohol. Slightly soluble in diethyl ether. Insoluble in benzene.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
109-57-9 | |
116.182 | |
HTKFORQRBXIQHD-UHFFFAOYSA-N | |
1549517 | |
prop-2-enylthiourea |
C4H8N2S | |
MFCD00004940 | |
allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin | |
CHEBI:74079 | |
C=CCNC(=S)N |
Specifications
109-57-9 | |
1.22 | |
C4H8N2S | |
25 g | |
1071470 | |
allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin | |
HTKFORQRBXIQHD-UHFFFAOYSA-N | |
prop-2-enylthiourea | |
1549517 | |
116.19 | |
N-Allylthiourea |
70°C to 78°C | |
Unpleasant | |
MFCD00004940 | |
UN2811 | |
14,9362 | |
Soluble in water and alcohol. Slightly soluble in diethyl ether. Insoluble in benzene. | |
C=CCNC(=S)N | |
116.182 | |
CHEBI:74079 | |
98% |
Safety and Handling
GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 203-683-5
RTECSNumber : YR8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only