Learn More
n-Amyl acetate, 99%
n-Amyl acetate, 99%, C7H14O2, CAS Number-628-63-7, 1-pentyl acetate, acetic acid, pentyl ester, amyl acetate, amyl acetic ester, amyl acetic ether, birnenoel, n-amyl acetate, n-pentyl acetate, n-pentyl ethanoate, pent-acetate, 2.5L, 98.5% min. (GC), 02, 131, 149.0 deg.C, 100.0, CHEBI:87362 | CAS: 628-63-7 | C7H14O2 | 130.19 g/mol
$129.72 - $359.52
Chemical Identifiers
| CAS | 628-63-7 |
|---|---|
| Molecular Formula | C7H14O2 |
| Molecular Weight (g/mol) | 130.19 |
| MDL Number | MFCD00009500 |
| InChI Key | PGMYKACGEOXYJE-UHFFFAOYSA-N |
| Synonym | amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether |
| PubChem CID | 12348 |
| ChEBI | CHEBI:87362 |
| IUPAC Name | pentyl acetate |
| SMILES | CCCCCOC(=O)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC149182500
|
Thermo Scientific Chemicals
149182500 |
250 mL | Glass bottle |
Each for $129.72
|
|
||||
|
AC149180010
|
Thermo Scientific Chemicals
149180010 |
1 L | Glass bottle |
Each for $307.96
|
|
||||
|
AC149180025
|
Thermo Scientific Chemicals
149180025 |
2.5 L | Glass bottle |
Each for $359.52
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 628-63-7 | |
| 130.19 | |
| PGMYKACGEOXYJE-UHFFFAOYSA-N | |
| 12348 | |
| pentyl acetate |
| C7H14O2 | |
| MFCD00009500 | |
| amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether | |
| CHEBI:87362 | |
| CCCCCOC(=O)C |
Specifications
| 628-63-7 | |
| 100.0 | |
| Colorless | |
| 149.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4010 to 1.4030 | |
| MFCD00009500 | |
| 02, 131 | |
| amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether | |
| PGMYKACGEOXYJE-UHFFFAOYSA-N | |
| pentyl acetate | |
| 12348 | |
| 130.19 | |
| Liquid |
| 98.5 | |
| -70.8°C | |
| 0.8700g/mL | |
| 24°C | |
| 98.5% min. (GC) | |
| C7H14O2 | |
| CH3CO2(CH2)4CH3 | |
| 250 mL | |
| 0.87 | |
| Solubility in water: 10g/L (20c). Other solubilities: miscible with alcohol and ether | |
| CCCCCOC(=O)C | |
| 130.19 | |
| CHEBI:87362 | |
| 99% | |
| n-Amyl acetate |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 211-047-3
RUO – Research Use Only