missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzoyl-L-tyrosine ethyl ester, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6343406

 View more versions of this product

Catalog No. AAH6343406


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

118°C to 121°C
3483-82-7
MFCD00002388
n-benzoyl-l-tyrosine ethyl ester, bz-tyr-oet, ethyl n-benzoyl-l-tyrosinate, btee, ethyl benzoyltyrosinate, benzoyltyrosine ethyl ester, benzoyl-l-tyrosine ethyl ester, s-ethyl 2-benzamido-3-4-hydroxyphenyl propanoate, benzoyl-tyr-oet, l-tyrosine, n-benzoyl-, ethyl ester
CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1
5 g
77033
98%
N-Benzoyl-L-tyrosine ethyl ester
C18H19NO4
2223819
SRLROPAFMUDDRC-XISACWJONA-N
ethyl (2S)-2-benzamido-3-(4-hydroxyphenyl)propanoate
313.35
313.35
Solid

Chemical Identifiers

3483-82-7
313.35
SRLROPAFMUDDRC-XISACWJONA-N
77033
CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1
C18H19NO4
MFCD00002388
n-benzoyl-l-tyrosine ethyl ester, bz-tyr-oet, ethyl n-benzoyl-l-tyrosinate, btee, ethyl benzoyltyrosinate, benzoyltyrosine ethyl ester, benzoyl-l-tyrosine ethyl ester, s-ethyl 2-benzamido-3-4-hydroxyphenyl propanoate, benzoyl-tyr-oet, l-tyrosine, n-benzoyl-, ethyl ester
ethyl (2S)-2-benzamido-3-(4-hydroxyphenyl)propanoate
Safety and Handling

Safety and Handling

EINECSNumber : 222-469-2

TSCA : Yes

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only