missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Benzyl-4-methoxybenzylamine, 97%, Thermo Scientific Chemicals

$832.25 - $832.25

Chemical Identifiers

CAS 14429-02-8
Molecular Formula C15H17NO
Molecular Weight (g/mol) 227.31
MDL Number MFCD01002920
InChI Key WZUBICZIDUTTNJ-UHFFFAOYSA-N
Synonym benzyl 4-methoxyphenyl methyl amine, benzyl-4-methoxy-benzyl-amine, n-benzyl-1-4-methoxyphenyl methanamine, benzyl-4-methoxybenzyl amine, n-benzyl-4-methoxybenzylamine, n-benzyl-n-4-methoxybenzyl amine, 4-methoxyphenyl methyl benzylamine, 4-methoxybenzylbenzylamine, cbmicro_025161, cbmicro_047998
PubChem CID 738564
IUPAC Name N-[(4-methoxyphenyl)methyl]-1-phenylmethanamine
SMILES COC1=CC=C(CNCC2=CC=CC=C2)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5563303
View Documents
Thermo Scientific Chemicals
H5563303
1 g
Each for $832.25
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14429-02-8
227.31
WZUBICZIDUTTNJ-UHFFFAOYSA-N
738564
COC1=CC=C(CNCC2=CC=CC=C2)C=C1
C15H17NO
MFCD01002920
benzyl 4-methoxyphenyl methyl amine, benzyl-4-methoxy-benzyl-amine, n-benzyl-1-4-methoxyphenyl methanamine, benzyl-4-methoxybenzyl amine, n-benzyl-4-methoxybenzylamine, n-benzyl-n-4-methoxybenzyl amine, 4-methoxyphenyl methyl benzylamine, 4-methoxybenzylbenzylamine, cbmicro_025161, cbmicro_047998
N-[(4-methoxyphenyl)methyl]-1-phenylmethanamine

Specifications

14429-02-8
MFCD01002920
UN3259
WZUBICZIDUTTNJ-UHFFFAOYSA-N
N-[(4-methoxyphenyl)methyl]-1-phenylmethanamine
738564
97%
C15H17NO
1 g
benzyl 4-methoxyphenyl methyl amine, benzyl-4-methoxy-benzyl-amine, n-benzyl-1-4-methoxyphenyl methanamine, benzyl-4-methoxybenzyl amine, n-benzyl-4-methoxybenzylamine, n-benzyl-n-4-methoxybenzyl amine, 4-methoxyphenyl methyl benzylamine, 4-methoxybenzylbenzylamine, cbmicro_025161, cbmicro_047998
COC1=CC=C(CNCC2=CC=CC=C2)C=C1
227.31
227.31
N-Benzyl-4-methoxybenzylamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only