missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Benzylacetamide 98.0+%, TCI America™
Supplier: TCI America B0404500G
Specifications
N-Benzylacetamide | |
61°C | |
157°C | |
MFCD00059204 | |
N-Acetylbenzylamine | |
CC(=O)NCC1=CC=CC=C1 | |
149.193 | |
149.19 | |
Crystalline Powder |
588-46-5 | |
White-Yellow | |
C9H11NO | |
500 g | |
UZJLYRRDVFWSGA-UHFFFAOYSA-N | |
N-benzylacetamide | |
11500 | |
≥98.0% (GC) |
Chemical Identifiers
588-46-5 | |
149.193 | |
UZJLYRRDVFWSGA-UHFFFAOYSA-N | |
11500 | |
CC(=O)NCC1=CC=CC=C1 |
C9H11NO | |
MFCD00059204 | |
N-Acetylbenzylamine | |
N-benzylacetamide |
Safety and Handling
EINECSNumber : (3)-0373&(9)-0024
RTECSNumber : AB4546300
TSCA : No