missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-D-glutamic-acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals

$175.64 - $175.64

Chemical Identifiers

CAS 51644-83-8
Molecular Formula C17H23NO6
Molecular Weight (g/mol) 337.372
MDL Number MFCD00077018
InChI Key GLMODRZPPBZPPB-CYBMUJFWSA-N
Synonym z-d-glu otbu-oh, z-d-glu obut-oh, d-glutamic acid,n-phenylmethoxy carbonyl-, 5-1,1-dimethylethyl ester, n-benzyloxy carbonyl-d-glutamic acid 5-tert-butyl ester, n-alpha-benzyloxycarbonyl-d-glutamic-acid-gamma-t-butyl ester, n-benzyloxycarbonyl-d-glutamic-acid 5-tert-butyl ester, r-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, 2r-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid
PubChem CID 6992708
IUPAC Name (2R)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6337106
View Documents
Thermo Scientific Chemicals
H6337106
5 g
Each for $175.64
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

51644-83-8
337.372
GLMODRZPPBZPPB-CYBMUJFWSA-N
6992708
CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
C17H23NO6
MFCD00077018
z-d-glu otbu-oh, z-d-glu obut-oh, d-glutamic acid,n-phenylmethoxy carbonyl-, 5-1,1-dimethylethyl ester, n-benzyloxy carbonyl-d-glutamic acid 5-tert-butyl ester, n-alpha-benzyloxycarbonyl-d-glutamic-acid-gamma-t-butyl ester, n-benzyloxycarbonyl-d-glutamic-acid 5-tert-butyl ester, r-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, 2r-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid
(2R)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid

Specifications

51644-83-8
MFCD00077018
z-d-glu otbu-oh, z-d-glu obut-oh, d-glutamic acid,n-phenylmethoxy carbonyl-, 5-1,1-dimethylethyl ester, n-benzyloxy carbonyl-d-glutamic acid 5-tert-butyl ester, n-alpha-benzyloxycarbonyl-d-glutamic-acid-gamma-t-butyl ester, n-benzyloxycarbonyl-d-glutamic-acid 5-tert-butyl ester, r-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, 2r-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid
CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
337.372
337.37
Solid
C17H23NO6
5 g
GLMODRZPPBZPPB-CYBMUJFWSA-N
(2R)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
6992708
98%
N-Benzyloxycarbonyl-D-glutamic-acid 5-tert-butyl ester
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only