missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine, 98%
CAS: 5545-54-0 | C21H25NO5 | 371.43 g/mol
Supplier: Thermo Scientific Chemicals H6676222
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine | |
| White | |
| MFCD00065701 | |
| z-tyr tbu-oh, s-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, n-cbz-o-tert-butyl-l-tyrosine, l-tyrosine, o-1,1-dimethylethyl-n-phenylmethoxy carbonyl, 2s-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, z-tyr but-oh, z-l-tyr tbu-oh, z-tyr t-bu-oh, n-benzyloxycarbonyl-o-t-butyl-l-tyrosine, o-tert-butyl-n-benzyloxycarbonyl tyrosine | |
| CC(C)(C)OC1=CC=C(CC(NC(=O)OCC2=CC=CC=C2)C(O)=O)C=C1 | |
| 100 g | |
| 978520 | |
| 98% |
| 5545-54-0 | |
| C21H25NO5 | |
| 2781067 | |
| YKVBQSGNGCKQSV-UHFFFAOYNA-N | |
| 2-{[(benzyloxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid | |
| 371.43 | |
| 371.43 | |
| Powder |
Chemical Identifiers
| 5545-54-0 | |
| 371.43 | |
| YKVBQSGNGCKQSV-UHFFFAOYNA-N | |
| 978520 | |
| CC(C)(C)OC1=CC=C(CC(NC(=O)OCC2=CC=CC=C2)C(O)=O)C=C1 |
| C21H25NO5 | |
| MFCD00065701 | |
| z-tyr tbu-oh, s-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, n-cbz-o-tert-butyl-l-tyrosine, l-tyrosine, o-1,1-dimethylethyl-n-phenylmethoxy carbonyl, 2s-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, z-tyr but-oh, z-l-tyr tbu-oh, z-tyr t-bu-oh, n-benzyloxycarbonyl-o-t-butyl-l-tyrosine, o-tert-butyl-n-benzyloxycarbonyl tyrosine | |
| 2-{[(benzyloxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only