missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-1,6-diaminohexane hydrochloride, 98%
CAS: 65915-94-8 | C11H25ClN2O2 | 252.78 g/mol
$104.15 - $360.12
Chemical Identifiers
| CAS | 65915-94-8 |
|---|---|
| Molecular Formula | C11H25ClN2O2 |
| Molecular Weight (g/mol) | 252.78 |
| MDL Number | MFCD00039072 |
| InChI Key | JSBWQIZQJOQPFN-UHFFFAOYSA-N |
| Synonym | n-boc-1,6-hexanediamine hydrochloride, tert-butyl 6-aminohexyl carbamate hydrochloride, n-boc-1,6-diamino-hexane hydrochloride, tert-butyl n-6-aminohexyl carbamate hydrochloride, n-6-aminohexyl tert-butoxy carboxamide, chloride, tert-butyl 6-aminohexyl carbamate monohydrochloride, tert-butyl 6-aminohexyl carbamate hcl, acmc-1b5cm, n-1-boc-1,6-diaminohexane . hcl, n-boc-1,6-hexanediamine hcl |
| PubChem CID | 3017638 |
| IUPAC Name | tert-butyl N-(6-aminohexyl)carbamate;hydrochloride |
| SMILES | [H+].[Cl-].CC(C)(C)OC(=O)NCCCCCCN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1948703
|
Thermo Scientific Chemicals
L1948703 |
1 g |
N/A
|
|
|||||
|
AAL1948706
|
Thermo Scientific Chemicals
L1948706 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 65915-94-8 | |
| 252.78 | |
| JSBWQIZQJOQPFN-UHFFFAOYSA-N | |
| 3017638 | |
| [H+].[Cl-].CC(C)(C)OC(=O)NCCCCCCN |
| C11H25ClN2O2 | |
| MFCD00039072 | |
| n-boc-1,6-hexanediamine hydrochloride, tert-butyl 6-aminohexyl carbamate hydrochloride, n-boc-1,6-diamino-hexane hydrochloride, tert-butyl n-6-aminohexyl carbamate hydrochloride, n-6-aminohexyl tert-butoxy carboxamide, chloride, tert-butyl 6-aminohexyl carbamate monohydrochloride, tert-butyl 6-aminohexyl carbamate hcl, acmc-1b5cm, n-1-boc-1,6-diaminohexane . hcl, n-boc-1,6-hexanediamine hcl | |
| tert-butyl N-(6-aminohexyl)carbamate;hydrochloride |
Specifications
| ∽164°C (decomposition) | |
| C11H25ClN2O2 | |
| 3631740 | |
| JSBWQIZQJOQPFN-UHFFFAOYSA-N | |
| tert-butyl N-(6-aminohexyl)carbamate;hydrochloride | |
| 252.78 | |
| 252.79 | |
| 98% |
| 65915-94-8 | |
| MFCD00039072 | |
| n-boc-1,6-hexanediamine hydrochloride, tert-butyl 6-aminohexyl carbamate hydrochloride, n-boc-1,6-diamino-hexane hydrochloride, tert-butyl n-6-aminohexyl carbamate hydrochloride, n-6-aminohexyl tert-butoxy carboxamide, chloride, tert-butyl 6-aminohexyl carbamate monohydrochloride, tert-butyl 6-aminohexyl carbamate hcl, acmc-1b5cm, n-1-boc-1,6-diaminohexane . hcl, n-boc-1,6-hexanediamine hcl | |
| [H+].[Cl-].CC(C)(C)OC(=O)NCCCCCCN | |
| 1 g | |
| 3017638 | |
| Hygroscopic | |
| N-Boc-1,6-diaminohexane hydrochloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 265-976-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only