missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-2-methyl-D-serine methyl ester, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H52421MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Boc-2-methyl-D-serine methyl ester | |
C10H19NO5 | |
250 mg | |
OUUNEDPIBZNRMT-SNVBAGLBSA-N | |
methyl (2R)-3-hydroxy-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate | |
10752097 | |
97% |
188476-33-7 | |
MFCD06797553 | |
n-boc-alpha-methyl-d-serine methyl ester, n-boc-2-methyl-d-serine methyl ester, n-tert-butoxycarbonyl-2-methyl-d-serine methyl ester, r-methyl 2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoate, methyl 2r-2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoate | |
CC(C)(C)OC(=O)NC(C)(CO)C(=O)OC | |
233.264 | |
233.26 |
Chemical Identifiers
188476-33-7 | |
233.264 | |
OUUNEDPIBZNRMT-SNVBAGLBSA-N | |
10752097 | |
CC(C)(C)OC(=O)NC(C)(CO)C(=O)OC |
C10H19NO5 | |
MFCD06797553 | |
n-boc-alpha-methyl-d-serine methyl ester, n-boc-2-methyl-d-serine methyl ester, n-tert-butoxycarbonyl-2-methyl-d-serine methyl ester, r-methyl 2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoate, methyl 2r-2-tert-butoxycarbonyl amino-3-hydroxy-2-methylpropanoate | |
methyl (2R)-3-hydroxy-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only