missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-3,3-diphenyl-L-alanine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5206306
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Boc-3,3-diphenyl-L-alanine | |
160°C (decomposition) | |
MFCD00191186 | |
4880641 | |
TYJDOLCFYZSNQC-KRWDZBQOSA-N | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3,3-diphenylpropanoic acid | |
2761487 | |
95% |
138662-63-2 | |
C20H23NO4 | |
5 g | |
boc-l-3,3-diphenylalanine, boc-3,3-diphenyl-l-alanine, s-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, s-n-boc-2-amino-3,3-diphenylpropionic acid, n-boc-beta-phenyl-l-phenylalanine, boc-beta-phenyl-phe-oh, n-tert-butoxycarbonyl-beta-phenyl-l-phenylalanine, s-2-tert-butoxycarbonylamino-3,3-diphenylpropanoic acid, 2s-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, pubchem20362 | |
CC(C)(C)OC(=O)NC(C(C1=CC=CC=C1)C2=CC=CC=C2)C(=O)O | |
341.407 | |
341.41 |
Chemical Identifiers
138662-63-2 | |
341.407 | |
TYJDOLCFYZSNQC-KRWDZBQOSA-N | |
2761487 | |
CC(C)(C)OC(=O)NC(C(C1=CC=CC=C1)C2=CC=CC=C2)C(=O)O |
C20H23NO4 | |
MFCD00191186 | |
boc-l-3,3-diphenylalanine, boc-3,3-diphenyl-l-alanine, s-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, s-n-boc-2-amino-3,3-diphenylpropionic acid, n-boc-beta-phenyl-l-phenylalanine, boc-beta-phenyl-phe-oh, n-tert-butoxycarbonyl-beta-phenyl-l-phenylalanine, s-2-tert-butoxycarbonylamino-3,3-diphenylpropanoic acid, 2s-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, pubchem20362 | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3,3-diphenylpropanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only