missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Boc-3,4-difluoro-D-phenylalanine, 98%, Thermo Scientific™

$93.19 - $93.19

Chemical Identifiers

CAS 205445-51-8
Molecular Formula C14H17F2NO4
Molecular Weight (g/mol) 301.29
MDL Number MFCD00672519
InChI Key CYAOPVHXASZUDE-LLVKDONJSA-N
Synonym boc-d-3,4-difluorophenylalanine, boc-3,4-difluoro-d-phenylalanine, boc-d-phe 3,4-dif-oh, boc-d-3,4-difluorophe, boc-d-phe 3,4-f2-oh, n-tert-butoxycarbonyl-3,4-difluoro-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-3,4-difluorophenyl propanoic acid, n-boc-3,4-difluoro-d-phenylalanine, n-alpha-t-butyloxycarbonyl-d-3,4-difluorophenylalanine, 3,4-difluoro-d-phenylalanine, boc protected
PubChem CID 2736194
IUPAC Name (2R)-3-(3,4-difluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1)F)F)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6637503
View Documents
Thermo Scientific Chemicals
H6637503
1 g
Each for $93.19
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

205445-51-8
301.29
CYAOPVHXASZUDE-LLVKDONJSA-N
2736194
CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1)F)F)C(=O)O
C14H17F2NO4
MFCD00672519
boc-d-3,4-difluorophenylalanine, boc-3,4-difluoro-d-phenylalanine, boc-d-phe 3,4-dif-oh, boc-d-3,4-difluorophe, boc-d-phe 3,4-f2-oh, n-tert-butoxycarbonyl-3,4-difluoro-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-3,4-difluorophenyl propanoic acid, n-boc-3,4-difluoro-d-phenylalanine, n-alpha-t-butyloxycarbonyl-d-3,4-difluorophenylalanine, 3,4-difluoro-d-phenylalanine, boc protected
(2R)-3-(3,4-difluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid

Specifications

205445-51-8
C14H17F2NO4
1920842
CYAOPVHXASZUDE-LLVKDONJSA-N
(2R)-3-(3,4-difluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
301.29
301.29
Powder
White
MFCD00672519
boc-d-3,4-difluorophenylalanine, boc-3,4-difluoro-d-phenylalanine, boc-d-phe 3,4-dif-oh, boc-d-3,4-difluorophe, boc-d-phe 3,4-f2-oh, n-tert-butoxycarbonyl-3,4-difluoro-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-3,4-difluorophenyl propanoic acid, n-boc-3,4-difluoro-d-phenylalanine, n-alpha-t-butyloxycarbonyl-d-3,4-difluorophenylalanine, 3,4-difluoro-d-phenylalanine, boc protected
CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1)F)F)C(=O)O
1 g
2736194
98%
N-Boc-3,4-difluoro-D-phenylalanine
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'