missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-3-methyl-L-phenylalanine, 98%, Thermo Scientific Chemicals
$161.19 - $629.37
Chemical Identifiers
CAS | 114873-06-2 |
---|---|
Molecular Formula | C15H21NO4 |
Molecular Weight (g/mol) | 279.336 |
MDL Number | MFCD01862944 |
InChI Key | HBBXWMALJNZDOM-LBPRGKRZSA-N |
Synonym | boc-phe 3-me-oh, boc-3-methyl-l-phenylalanine, boc-l-3-methylphenylalanine, boc-l-3-methylphe, boc-3-methy-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-m-tolyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-methylphenyl propanoic acid, boc-l-3-methyl phenylalanine, boc-phe 3-me-oh tlc, 3-methyl-n-t-butoxycarbonyl-l-phenylalanine |
PubChem CID | 2761470 |
IUPAC Name | (2S)-3-(3-methylphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
SMILES | CC1=CC(=CC=C1)CC(C(=O)O)NC(=O)OC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6286003
|
Thermo Scientific Chemicals
H6286003 |
1 g |
Each for $161.19
|
|
|||||
AAH6286006
|
Thermo Scientific Chemicals
H6286006 |
5 g |
Each for $629.37
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
114873-06-2 | |
279.336 | |
HBBXWMALJNZDOM-LBPRGKRZSA-N | |
2761470 | |
CC1=CC(=CC=C1)CC(C(=O)O)NC(=O)OC(C)(C)C |
C15H21NO4 | |
MFCD01862944 | |
boc-phe 3-me-oh, boc-3-methyl-l-phenylalanine, boc-l-3-methylphenylalanine, boc-l-3-methylphe, boc-3-methy-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-m-tolyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-methylphenyl propanoic acid, boc-l-3-methyl phenylalanine, boc-phe 3-me-oh tlc, 3-methyl-n-t-butoxycarbonyl-l-phenylalanine | |
(2S)-3-(3-methylphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
114873-06-2 | |
MFCD01862944 | |
boc-phe 3-me-oh, boc-3-methyl-l-phenylalanine, boc-l-3-methylphenylalanine, boc-l-3-methylphe, boc-3-methy-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-m-tolyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-methylphenyl propanoic acid, boc-l-3-methyl phenylalanine, boc-phe 3-me-oh tlc, 3-methyl-n-t-butoxycarbonyl-l-phenylalanine | |
CC1=CC(=CC=C1)CC(C(=O)O)NC(=O)OC(C)(C)C | |
279.336 | |
279.33 | |
1 g | |
N-Boc-3-methyl-L-phenylalanine |
C15H21NO4 | |
8989098 | |
HBBXWMALJNZDOM-LBPRGKRZSA-N | |
(2S)-3-(3-methylphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
2761470 | |
98% | |
Solid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only