missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-beta-phenyl-D-phenylalanine, 98%
CAS: 143060-31-5 | C20H23NO4 | 341.41 g/mol
$296.58 - $501.34
Chemical Identifiers
| CAS | 143060-31-5 |
|---|---|
| Molecular Formula | C20H23NO4 |
| Molecular Weight (g/mol) | 341.41 |
| MDL Number | MFCD00191187 |
| InChI Key | TYJDOLCFYZSNQC-UHFFFAOYNA-N |
| Synonym | boc-d-3,3-diphenylalanine, boc-d-ala 3,3-diphenyl-oh, r-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, boc-3,3-diphenyl-d-alanine, r-n-boc-2-amino-3,3-diphenylpropionic acid, n-boc-beta-phenyl-d-phenylalanine, n-tert-butoxycarbonyl-beta-phenyl-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, n-tert-butoxycarbonyl-3-phenyl-l-phenylalanine, n-boc-b-phenyl-d-phenylalanine |
| PubChem CID | 7019136 |
| SMILES | CC(C)(C)OC(=O)NC(C(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH56885ME
|
Thermo Scientific Chemicals
H56885ME |
500 mg |
Each for $296.58
|
|
|||||
|
AAH5688503
|
Thermo Scientific Chemicals
H5688503 |
1 g |
Each for $501.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 143060-31-5 | |
| 341.41 | |
| TYJDOLCFYZSNQC-UHFFFAOYNA-N | |
| 7019136 |
| C20H23NO4 | |
| MFCD00191187 | |
| boc-d-3,3-diphenylalanine, boc-d-ala 3,3-diphenyl-oh, r-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, boc-3,3-diphenyl-d-alanine, r-n-boc-2-amino-3,3-diphenylpropionic acid, n-boc-beta-phenyl-d-phenylalanine, n-tert-butoxycarbonyl-beta-phenyl-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, n-tert-butoxycarbonyl-3-phenyl-l-phenylalanine, n-boc-b-phenyl-d-phenylalanine | |
| CC(C)(C)OC(=O)NC(C(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O |
Specifications
| 143060-31-5 | |
| C20H23NO4 | |
| 500 mg | |
| TYJDOLCFYZSNQC-UHFFFAOYNA-N | |
| CC(C)(C)OC(=O)NC(C(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O | |
| 341.41 | |
| 341.4 | |
| N-Boc-beta-phenyl-D-phenylalanine |
| 157°C | |
| MFCD00191187 | |
| boc-d-3,3-diphenylalanine, boc-d-ala 3,3-diphenyl-oh, r-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, boc-3,3-diphenyl-d-alanine, r-n-boc-2-amino-3,3-diphenylpropionic acid, n-boc-beta-phenyl-d-phenylalanine, n-tert-butoxycarbonyl-beta-phenyl-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, n-tert-butoxycarbonyl-3-phenyl-l-phenylalanine, n-boc-b-phenyl-d-phenylalanine | |
| −30 (c=1 in Ethanol) | |
| 2-{[(tert-butoxy)carbonyl]amino}-3,3-diphenylpropanoic acid | |
| 7019136 | |
| 98% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only