missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-D-glutamic acid, 98%, Thermo Scientific Chemicals
$161.68 - $691.90
Chemical Identifiers
CAS | 34404-28-9 |
---|---|
Molecular Formula | C10H17NO6 |
Molecular Weight (g/mol) | 247.247 |
MDL Number | MFCD00190790 |
InChI Key | AQTUACKQXJNHFQ-ZCFIWIBFSA-N |
Synonym | boc-d-glu-oh, boc-d-glutamic acid, n-tert-butoxycarbonyl-d-glutamic acid, r-2-tert-butoxycarbonyl amino pentanedioic acid, d-glutamic acid, n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino pentanedioic acid, pubchem20973, n-boc-d-glutamic acid, ksc493m2t, 2r-2-2-methylpropan-2-yl oxycarbonylamino pentanedioic acid |
PubChem CID | 7019046 |
IUPAC Name | (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioic acid |
SMILES | CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6276206
|
Thermo Scientific Chemicals
H6276206 |
5 g |
Each for $161.68
|
|
|||||
AAH6276214
|
Thermo Scientific Chemicals
H6276214 |
25 g |
Each for $691.90
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
34404-28-9 | |
247.247 | |
AQTUACKQXJNHFQ-ZCFIWIBFSA-N | |
7019046 | |
CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)O |
C10H17NO6 | |
MFCD00190790 | |
boc-d-glu-oh, boc-d-glutamic acid, n-tert-butoxycarbonyl-d-glutamic acid, r-2-tert-butoxycarbonyl amino pentanedioic acid, d-glutamic acid, n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino pentanedioic acid, pubchem20973, n-boc-d-glutamic acid, ksc493m2t, 2r-2-2-methylpropan-2-yl oxycarbonylamino pentanedioic acid | |
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioic acid |
Specifications
34404-28-9 | |
MFCD00190790 | |
boc-d-glu-oh, boc-d-glutamic acid, n-tert-butoxycarbonyl-d-glutamic acid, r-2-tert-butoxycarbonyl amino pentanedioic acid, d-glutamic acid, n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino pentanedioic acid, pubchem20973, n-boc-d-glutamic acid, ksc493m2t, 2r-2-2-methylpropan-2-yl oxycarbonylamino pentanedioic acid | |
CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)O | |
247.247 | |
247.25 | |
N-Boc-D-glutamic acid |
C10H17NO6 | |
5 g | |
AQTUACKQXJNHFQ-ZCFIWIBFSA-N | |
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioic acid | |
7019046 | |
98% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only