missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Boc-D-tyrosine methyl ester, 96%, Thermo Scientific Chemicals

$84.30 - $301.73

Chemical Identifiers

CAS 76757-90-9
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.34
MDL Number MFCD00057824
InChI Key NQIFXJSLCUJHBB-UHFFFAOYNA-N
Synonym boc-d-tyr-ome, r-methyl 2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoate, boc-d-tyrosine methyl ester, d-tyrosine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoate, methyl 2r-2-tert-butoxy carbonyl amino-3-4-hydroxyphenyl propanoate, methyl boc-d-tyrosinate, boc-d-tyrosine-methyl ester, n-t-butoxycarbonyl-d-tyrosine methyl ester
PubChem CID 7016055
IUPAC Name methyl 2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
SMILES COC(=O)C(CC1=CC=C(O)C=C1)NC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6682706
View Documents Promotion Details
Thermo Scientific Chemicals
H6682706
5 g N/A
 
AAH6682714
View Documents Promotion Details
Thermo Scientific Chemicals
H6682714
25 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

76757-90-9
295.34
NQIFXJSLCUJHBB-UHFFFAOYNA-N
7016055
COC(=O)C(CC1=CC=C(O)C=C1)NC(=O)OC(C)(C)C
C15H21NO5
MFCD00057824
boc-d-tyr-ome, r-methyl 2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoate, boc-d-tyrosine methyl ester, d-tyrosine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoate, methyl 2r-2-tert-butoxy carbonyl amino-3-4-hydroxyphenyl propanoate, methyl boc-d-tyrosinate, boc-d-tyrosine-methyl ester, n-t-butoxycarbonyl-d-tyrosine methyl ester
methyl 2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate

Specifications

76757-90-9
C15H21NO5
4704567
NQIFXJSLCUJHBB-UHFFFAOYNA-N
methyl 2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
7016055
96%
Powder
White
MFCD00057824
boc-d-tyr-ome, r-methyl 2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoate, boc-d-tyrosine methyl ester, d-tyrosine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2r-2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoate, methyl 2r-2-tert-butoxy carbonyl amino-3-4-hydroxyphenyl propanoate, methyl boc-d-tyrosinate, boc-d-tyrosine-methyl ester, n-t-butoxycarbonyl-d-tyrosine methyl ester
COC(=O)C(CC1=CC=C(O)C=C1)NC(=O)OC(C)(C)C
295.34
295.34
5 g
N-Boc-D-tyrosine methyl ester
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only