missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-D-valine, 98+%, Thermo Scientific Chemicals
$52.22 - $164.73
Chemical Identifiers
CAS | 22838-58-0 |
---|---|
Molecular Formula | C10H19NO4 |
Molecular Weight (g/mol) | 217.27 |
MDL Number | MFCD00038282 |
InChI Key | SZXBQTSZISFIAO-SSDOTTSWSA-N |
Synonym | boc-d-valine, boc-d-val-oh, n-boc-d-valine, n-tert-butoxycarbonyl-d-valine, tert-butoxycarbonyl-d-valine, r-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, d-valine, n-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-valin, r-2-tert-butoxycarbonylamino-3-methylbutanoic acid, n-alpha-t-butyloxycarbonyl-d-valine |
PubChem CID | 637605 |
IUPAC Name | (2R)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
SMILES | CC(C)[C@@H](NC(=O)OC(C)(C)C)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0919303
|
Thermo Scientific Chemicals
L0919303 |
1 g |
Each for $52.22
|
|
||||||
AAL0919306
|
Thermo Scientific Chemicals
L0919306 |
5 g |
Each for $164.73
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22838-58-0 | |
217.27 | |
SZXBQTSZISFIAO-SSDOTTSWSA-N | |
637605 | |
CC(C)[C@@H](NC(=O)OC(C)(C)C)C(O)=O |
C10H19NO4 | |
MFCD00038282 | |
boc-d-valine, boc-d-val-oh, n-boc-d-valine, n-tert-butoxycarbonyl-d-valine, tert-butoxycarbonyl-d-valine, r-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, d-valine, n-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-valin, r-2-tert-butoxycarbonylamino-3-methylbutanoic acid, n-alpha-t-butyloxycarbonyl-d-valine | |
(2R)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
Specifications
22838-58-0 | |
C10H19NO4 | |
1 g | |
boc-d-valine, boc-d-val-oh, n-boc-d-valine, n-tert-butoxycarbonyl-d-valine, tert-butoxycarbonyl-d-valine, r-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, d-valine, n-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-valin, r-2-tert-butoxycarbonylamino-3-methylbutanoic acid, n-alpha-t-butyloxycarbonyl-d-valine | |
+6.3° (c=1 in Acetic Acid) | |
(2R)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid | |
637605 | |
≥98% |
76°C to 80°C | |
MFCD00038282 | |
2050408 | |
SZXBQTSZISFIAO-SSDOTTSWSA-N | |
CC(C)[C@@H](NC(=O)OC(C)(C)C)C(O)=O | |
217.27 | |
217.27 | |
N-Boc-D-valine |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only