Learn More
N-Boc-ethylenediamine, 98%, may cont up to 5% tert-butanol
CAS: 57260-73-8 | C7H16N2O2 | 160.22 g/mol
$245.38 - $893.25
Chemical Identifiers
| CAS | 57260-73-8 |
|---|---|
| Molecular Formula | C7H16N2O2 |
| Molecular Weight (g/mol) | 160.22 |
| MDL Number | MFCD00191871 |
| InChI Key | AOCSUUGBCMTKJH-UHFFFAOYSA-N |
| Synonym | n-boc-ethylenediamine, tert-butyl n-2-aminoethyl carbamate, tert-butyl 2-aminoethyl carbamate, n-t-boc-ethylenediamine, n-tert-boc-ethylenediamine, tert-butyl 2-aminoethylcarbamate, n-boc-1,2-diaminoethane, n-boc-ethane-1,2-diamine, mono-n-t-boc-eda, n-boc-diaminoethane |
| PubChem CID | 187201 |
| IUPAC Name | tert-butyl N-(2-aminoethyl)carbamate |
| SMILES | CC(C)(C)OC(=O)NCCN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1994706
|
Thermo Scientific Chemicals
L1994706 |
5 g |
Each for $245.38
|
|
|||||
|
AAL1994714
|
Thermo Scientific Chemicals
L1994714 |
25 g |
Each for $893.25
|
|
|||||
Description
N-Boc-ethylenediamine is used in the synthesis of thyronamine derivatives. It is also used in the preparation of (2-isothiocyanato-ethyl)-carbamic acid tert-butyl ester by reacting with carbon disulfide. Further, it is used in both oligonucleotide and peptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-ethylenediamine is used in the synthesis of thyronamine derivatives. It is also used in the preparation of (2-isothiocyanato-ethyl)-carbamic acid tert-butyl ester by reacting with carbon disulfide. Further, it is used in both oligonucleotide and peptide synthesis.
Solubility
Miscible with methanol and chloroform. Slightly miscible with water.
Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents and acids.
Chemical Identifiers
| 57260-73-8 | |
| 160.22 | |
| AOCSUUGBCMTKJH-UHFFFAOYSA-N | |
| 187201 | |
| CC(C)(C)OC(=O)NCCN |
| C7H16N2O2 | |
| MFCD00191871 | |
| n-boc-ethylenediamine, tert-butyl n-2-aminoethyl carbamate, tert-butyl 2-aminoethyl carbamate, n-t-boc-ethylenediamine, n-tert-boc-ethylenediamine, tert-butyl 2-aminoethylcarbamate, n-boc-1,2-diaminoethane, n-boc-ethane-1,2-diamine, mono-n-t-boc-eda, n-boc-diaminoethane | |
| tert-butyl N-(2-aminoethyl)carbamate |
Specifications
| 57260-73-8 | |
| C7H16N2O2 | |
| MFCD00191871 | |
| 1932330 | |
| Miscible with methanol and chloroform. Slightly miscible with water. | |
| 1.016 | |
| tert-butyl N-(2-aminoethyl)carbamate | |
| 5 g | |
| 187201 | |
| 98% | |
| N-Boc-ethylenediamine |
| 72°C (0.1mmHg) | |
| 1.457 | |
| UN2735 | |
| n-boc-ethylenediamine, tert-butyl n-2-aminoethyl carbamate, tert-butyl 2-aminoethyl carbamate, n-t-boc-ethylenediamine, n-tert-boc-ethylenediamine, tert-butyl 2-aminoethylcarbamate, n-boc-1,2-diaminoethane, n-boc-ethane-1,2-diamine, mono-n-t-boc-eda, n-boc-diaminoethane | |
| AOCSUUGBCMTKJH-UHFFFAOYSA-N | |
| CC(C)(C)OC(=O)NCCN | |
| 160.22 | |
| >110°C (230°F) | |
| 160.22 | |
| Air Sensitive | |
| may cont up to 5% tert-butanol |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335-H336
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only