missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-glycine tert-butyl ester, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6237503
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Boc-glycine tert-butyl ester | |
66°C to 68°C | |
(CH3)3COCONHCH2CO2C(CH3)3 | |
1 g | |
boc-gly-otbu, tert-butyl 2-tert-butoxycarbonyl amino acetate, boc-glycine tert-butyl ester, n-boc-glycine tert-butyl ester, n-tert-butoxycarbonyl glycine tert-butyl ester, glycine, n-1,1-dimethylethoxy carbonyl-, 1,1-dimethylethyl ester, tert-butyl 2-2-methylpropan-2-yl oxycarbonylamino acetate, 2-azabutanedioic acid di-tert-butyl ester | |
CC(C)(C)OC(=O)CNC(=O)OC(C)(C)C | |
231.29 | |
231.29 |
111652-20-1 | |
C11H21NO4 | |
MFCD00191866 | |
5002580 | |
ZDKFMHKWZATBMR-UHFFFAOYSA-N | |
tert-butyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate | |
4097102 | |
95% |
Chemical Identifiers
111652-20-1 | |
231.29 | |
ZDKFMHKWZATBMR-UHFFFAOYSA-N | |
4097102 |
C11H21NO4 | |
MFCD00191866 | |
boc-gly-otbu, tert-butyl 2-tert-butoxycarbonyl amino acetate, boc-glycine tert-butyl ester, n-boc-glycine tert-butyl ester, n-tert-butoxycarbonyl glycine tert-butyl ester, glycine, n-1,1-dimethylethoxy carbonyl-, 1,1-dimethylethyl ester, tert-butyl 2-2-methylpropan-2-yl oxycarbonylamino acetate, 2-azabutanedioic acid di-tert-butyl ester | |
CC(C)(C)OC(=O)CNC(=O)OC(C)(C)C |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only