missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-alaninamide, 96%
CAS: 85642-13-3 | C8H16N2O3 | 188.23 g/mol
$103.44 - $1175.70
Chemical Identifiers
| CAS | 85642-13-3 |
|---|---|
| Molecular Formula | C8H16N2O3 |
| Molecular Weight (g/mol) | 188.23 |
| MDL Number | MFCD03701447 |
| InChI Key | GZKKSKKIABUUCX-UHFFFAOYNA-N |
| Synonym | boc-ala-nh2, n-t-butyloxycarbonyl-l-alanine amide, s-tert-butyl 1-amino-1-oxopropan-2-yl carbamate, tert-butyl n-1s-1-carbamoylethyl carbamate, boc-beta-ala-nh2, boc-l-ala-nh2, boc-l-alanine-nh2, boc-l-alanine amide, s-boc-alaninamide, ambotzbaa5310 |
| PubChem CID | 7015295 |
| SMILES | CC(NC(=O)OC(C)(C)C)C(N)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH62604MD
|
Thermo Scientific Chemicals
H62604MD |
250 mg |
Each for $103.44
|
|
|||||
|
AAH6260403
|
Thermo Scientific Chemicals
H6260403 |
1 g |
Each for $326.58
|
|
|||||
|
AAH6260406
|
Thermo Scientific Chemicals
H6260406 |
5 g |
Each for $1,175.70
|
|
|||||
Description
N-Boc-L-alaninamide is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-L-alaninamide is used as pharmaceutical intermediate.
Solubility
Soluble in water or 1% acetic acid.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
| 85642-13-3 | |
| 188.23 | |
| GZKKSKKIABUUCX-UHFFFAOYNA-N | |
| 7015295 |
| C8H16N2O3 | |
| MFCD03701447 | |
| boc-ala-nh2, n-t-butyloxycarbonyl-l-alanine amide, s-tert-butyl 1-amino-1-oxopropan-2-yl carbamate, tert-butyl n-1s-1-carbamoylethyl carbamate, boc-beta-ala-nh2, boc-l-ala-nh2, boc-l-alanine-nh2, boc-l-alanine amide, s-boc-alaninamide, ambotzbaa5310 | |
| CC(NC(=O)OC(C)(C)C)C(N)=O |
Specifications
| 85642-13-3 | |
| MFCD03701447 | |
| boc-ala-nh2, n-t-butyloxycarbonyl-l-alanine amide, s-tert-butyl 1-amino-1-oxopropan-2-yl carbamate, tert-butyl n-1s-1-carbamoylethyl carbamate, boc-beta-ala-nh2, boc-l-ala-nh2, boc-l-alanine-nh2, boc-l-alanine amide, s-boc-alaninamide, ambotzbaa5310 | |
| GZKKSKKIABUUCX-UHFFFAOYNA-N | |
| tert-butyl N-(1-carbamoylethyl)carbamate | |
| 7015295 | |
| 96% | |
| N-Boc-L-alaninamide |
| C8H16N2O3 | |
| 250 mg | |
| Soluble in water or 1% acetic acid. | |
| CC(NC(=O)OC(C)(C)C)C(N)=O | |
| 188.23 | |
| 188.22 | |
| Solid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only