missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-histidine methyl ester, 95%

CAS: 2488-14-4 | C12H19N3O4 | 269.301 g/mol

$154.47 - $545.48

Chemical Identifiers

CAS 2488-14-4
Molecular Formula C12H19N3O4
Molecular Weight (g/mol) 269.301
MDL Number MFCD00167635
InChI Key RWEVEEORUORBAX-VIFPVBQESA-N
Synonym boc-his-ome, nalpha-boc-l-histidine methyl ester, l-histidine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoate, n-boc-l-histidine methyl ester, na-boc-l-histidine methyl ester, n-tert-butoxycarbonyl-l-histidine methyl ester, l-histidine,n-1,1-dimethylethoxy carbonyl-, methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoate, s-methyl 2-tert-butoxycarbonyl amino-3-1h-imidazol-5-yl propanoate
PubChem CID 7017200
IUPAC Name methyl (2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6254503
View Documents
Thermo Scientific Chemicals
H6254503
1 g
Each for $154.47
Only null left
 
AAH6254506
View Documents
Thermo Scientific Chemicals
H6254506
5 g
Each for $545.48
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2488-14-4
269.301
RWEVEEORUORBAX-VIFPVBQESA-N
7017200
CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)OC
C12H19N3O4
MFCD00167635
boc-his-ome, nalpha-boc-l-histidine methyl ester, l-histidine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoate, n-boc-l-histidine methyl ester, na-boc-l-histidine methyl ester, n-tert-butoxycarbonyl-l-histidine methyl ester, l-histidine,n-1,1-dimethylethoxy carbonyl-, methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoate, s-methyl 2-tert-butoxycarbonyl amino-3-1h-imidazol-5-yl propanoate
methyl (2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate

Specifications

2488-14-4
MFCD00167635
RWEVEEORUORBAX-VIFPVBQESA-N
methyl (2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
269.301
269.3
Solid
C12H19N3O4
boc-his-ome, nalpha-boc-l-histidine methyl ester, l-histidine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoate, n-boc-l-histidine methyl ester, na-boc-l-histidine methyl ester, n-tert-butoxycarbonyl-l-histidine methyl ester, l-histidine,n-1,1-dimethylethoxy carbonyl-, methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-1h-imidazol-4-yl propanoate, s-methyl 2-tert-butoxycarbonyl amino-3-1h-imidazol-5-yl propanoate
CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)OC
1 g
7017200
95%
N-Boc-L-histidine methyl ester
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only