missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-L-isoleucine, 98+%, Thermo Scientific Chemicals
$64.08 - $199.05
Chemical Identifiers
CAS | 13139-16-7 |
---|---|
Molecular Formula | C11H21NO4 |
Molecular Weight (g/mol) | 231.29 |
MDL Number | MFCD00038324 MFCD00237550 |
InChI Key | QJCNLJWUIOIMMF-YUMQZZPRSA-N |
Synonym | boc-l-isoleucine, boc-ile-oh, n-boc-l-isoleucine, n-tert-butoxycarbonyl-l-isoleucine, boc-dl-ile-oh, boc-l-ile-oh, 2s,3s-2-tert-butoxycarbonyl amino-3-methylpentanoic acid, l-isoleucine, n-1,1-dimethylethoxy carbonyl, boc-isoleucine, n-t-boc-l-isoleucine |
PubChem CID | 2724762 |
IUPAC Name | (2S,3S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid |
SMILES | CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1602106
|
Thermo Scientific Chemicals
A1602106 |
5 g |
Each for $64.08
|
|
||||||
AAA1602114
|
Thermo Scientific Chemicals
A1602114 |
25 g |
Each for $199.05
|
|
||||||
Description
N-Boc-L-isoleucine is used to prepare BOC-L-isoleucine hydroxysuccinimide ester by using 1-hydroxybenzotriazole and 1-ethyl-3-<3-(dimethylamino)propyl>carbodiimide hydrochloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-L-isoleucine is used to prepare BOC-L-isoleucine hydroxysuccinimide ester by using 1-hydroxybenzotriazole and 1-ethyl-3-<3-(dimethylamino)propyl>carbodiimide hydrochloride.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases. Store in a cool place.
Chemical Identifiers
13139-16-7 | |
231.29 | |
QJCNLJWUIOIMMF-YUMQZZPRSA-N | |
2724762 | |
CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O |
C11H21NO4 | |
MFCD00038324 MFCD00237550 | |
boc-l-isoleucine, boc-ile-oh, n-boc-l-isoleucine, n-tert-butoxycarbonyl-l-isoleucine, boc-dl-ile-oh, boc-l-ile-oh, 2s,3s-2-tert-butoxycarbonyl amino-3-methylpentanoic acid, l-isoleucine, n-1,1-dimethylethoxy carbonyl, boc-isoleucine, n-t-boc-l-isoleucine | |
(2S,3S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid |
Specifications
13139-16-7 | |
C11H21NO4 | |
5 g | |
boc-l-isoleucine, boc-ile-oh, n-boc-l-isoleucine, n-tert-butoxycarbonyl-l-isoleucine, boc-dl-ile-oh, boc-l-ile-oh, 2s,3s-2-tert-butoxycarbonyl amino-3-methylpentanoic acid, l-isoleucine, n-1,1-dimethylethoxy carbonyl, boc-isoleucine, n-t-boc-l-isoleucine | |
QJCNLJWUIOIMMF-YUMQZZPRSA-N | |
(2S,3S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid | |
2724762 | |
≥98% |
67°C to 70°C | |
MFCD00038324 MFCD00237550 | |
1711700 | |
Soluble in methanol. Insoluble in water. | |
CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O | |
231.29 | |
231.06 | |
N-Boc-L-isoleucine |
Safety and Handling
EINECSNumber : 236-074-8
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only