missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-phenylalanine, 99%

CAS: 13734-34-4 | C14H19NO4 | 265.31 g/mol

$64.26 - $257.74

Chemical Identifiers

CAS 13734-34-4
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00002663
InChI Key ZYJPUMXJBDHSIF-NSHDSACASA-N
Synonym boc-phe-oh, boc-l-phenylalanine, boc-phenylalanine, n-tert-butoxycarbonyl-l-phenylalanine, n-boc-l-phenylalanine, n-t-boc-l-phenylalanine, n-alpha-t-boc-l-phenylalanine, n-t-butyloxycarbonyl-l-phenylalanine, l-phenylalanine, n-1,1-dimethylethoxy carbonyl, boc-phe
PubChem CID 77857
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1601706
View Documents
Thermo Scientific Chemicals
A1601706
5 g
Each for $64.26
Only null left
 
AAA1601714
View Documents
Thermo Scientific Chemicals
A1601714
25 g
Each for $257.74
Only null left
 
Description

Description

N-Boc-L-phenylalanine is a derivative of Phenylalanine used in enantioselective hydrolysis of amino acid esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-phenylalanine is a derivative of Phenylalanine used in enantioselective hydrolysis of amino acid esters.

Solubility
Soluble in methanol, dichloromethane, dimethylformamide and N-methyl-2-pyrrolidone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

13734-34-4
265.31
ZYJPUMXJBDHSIF-NSHDSACASA-N
77857
C14H19NO4
MFCD00002663
boc-phe-oh, boc-l-phenylalanine, boc-phenylalanine, n-tert-butoxycarbonyl-l-phenylalanine, n-boc-l-phenylalanine, n-t-boc-l-phenylalanine, n-alpha-t-boc-l-phenylalanine, n-t-butyloxycarbonyl-l-phenylalanine, l-phenylalanine, n-1,1-dimethylethoxy carbonyl, boc-phe
CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

13734-34-4
C14H19NO4
5 g
boc-phe-oh, boc-l-phenylalanine, boc-phenylalanine, n-tert-butoxycarbonyl-l-phenylalanine, n-boc-l-phenylalanine, n-t-boc-l-phenylalanine, n-alpha-t-boc-l-phenylalanine, n-t-butyloxycarbonyl-l-phenylalanine, l-phenylalanine, n-1,1-dimethylethoxy carbonyl, boc-phe
ZYJPUMXJBDHSIF-NSHDSACASA-N
CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
265.31
265.31
N-Boc-L-phenylalanine
84°C to 88°C
MFCD00002663
2219729
Soluble in methanol,dichloromethane,dimethylformamide and N-methyl-2-pyrrolidone.
+25° (c=1 in Ethanol)
2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoic acid
77857
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 237-305-5

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only