missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-benzyl-D-tyrosine, 95%

CAS: 63769-58-4 | C21H25NO5 | 371.43 g/mol

$187.12 - $624.92

Chemical Identifiers

CAS 63769-58-4
Molecular Formula C21H25NO5
Molecular Weight (g/mol) 371.43
MDL Number MFCD00038249
InChI Key ZAVSPTOJKOFMTA-LGWFVXIRNA-N
Synonym boc-d-tyr bzl-oh, boc-o-benzyl-d-tyrosine, n-boc-o-benzyl-d-tyrosine, boc-o-benzyl-d-tyr, r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, ambotzbaa1381, boc-dtyr bn-oh, pubchem14949, boc-d-tyr bzl oh
PubChem CID 6992575
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylmethoxyphenyl)propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6342706
View Documents
Thermo Scientific Chemicals
H6342706
5 g
Each for $187.12
Only null left
 
AAH6342714
View Documents
Thermo Scientific Chemicals
H6342714
25 g
Each for $624.92
Only null left
 
Description

Description

N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

63769-58-4
371.43
ZAVSPTOJKOFMTA-LGWFVXIRNA-N
6992575
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O
C21H25NO5
MFCD00038249
boc-d-tyr bzl-oh, boc-o-benzyl-d-tyrosine, n-boc-o-benzyl-d-tyrosine, boc-o-benzyl-d-tyr, r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, ambotzbaa1381, boc-dtyr bn-oh, pubchem14949, boc-d-tyr bzl oh
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylmethoxyphenyl)propanoic acid

Specifications

63769-58-4
MFCD00038249
Slightly soluble in water.
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O
5 g
6992575
95%
N-Boc-O-benzyl-D-tyrosine
C21H25NO5
boc-d-tyr bzl-oh, boc-o-benzyl-d-tyrosine, n-boc-o-benzyl-d-tyrosine, boc-o-benzyl-d-tyr, r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, ambotzbaa1381, boc-dtyr bn-oh, pubchem14949, boc-d-tyr bzl oh
ZAVSPTOJKOFMTA-LGWFVXIRNA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylmethoxyphenyl)propanoic acid
371.43
371.43
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only