Learn More
N-Bromosuccinimide, 99%
CAS: 128-08-5 | C4H4BrNO2 | 177.99 g/mol
$80.82 - $993.25
Chemical Identifiers
| CAS | 128-08-5 |
|---|---|
| MDL Number | MFCD00005510 |
| InChI Key | PCLIMKBDDGJMGD-UHFFFAOYSA-N |
| Synonym | n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide |
| PubChem CID | 67184 |
| ChEBI | CHEBI:53174 |
| IUPAC Name | 1-bromopyrrolidine-2,5-dione |
| SMILES | C1CC(=O)N(C1=O)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC107450100
|
Thermo Scientific Chemicals
107450100 |
10 g | Glass bottle |
Each for $80.82
|
|
||||
|
AC107451000
|
Thermo Scientific Chemicals
107451000 |
100 g | Glass bottle |
Each for $83.98
|
|
||||
|
AC107455000
|
Thermo Scientific Chemicals
107455000 |
500 g | Glass bottle |
Each for $185.77
|
|
||||
|
AC107450010
|
Thermo Scientific Chemicals
107450010 |
1 kg | Glass bottle |
Each for $283.22
|
|
||||
|
AC107450050
|
Thermo Scientific Chemicals
107450050 |
5 kg | Plastic drum |
Each for $993.25
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent, C-X bond formation reagentChemical Identifiers
| 128-08-5 | |
| PCLIMKBDDGJMGD-UHFFFAOYSA-N | |
| 67184 | |
| 1-bromopyrrolidine-2,5-dione |
| MFCD00005510 | |
| n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide | |
| CHEBI:53174 | |
| C1CC(=O)N(C1=O)Br |
Specifications
| 128-08-5 | |
| 100.0 | |
| White to Yellow | |
| 1% max. | |
| 98.5 to 101.5% (on dry substance) (Iodometry) | |
| C4H4BrNO2 | |
| 10 g | |
| 01,78; 02,40; 03,34; 04,49; 05,65; 06,74; 07,37; 08,54; 09,70; 10,57; 11,79; 12,79; 13,41; 14,57; 15,50; 16,49 | |
| n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide | |
| PCLIMKBDDGJMGD-UHFFFAOYSA-N | |
| 1-bromopyrrolidine-2,5-dione | |
| 67184 | |
| 177.99 | |
| Crystalline Powder |
| 98.5 | |
| 175.0°C to 178.0°C | |
| 180°C | |
| Authentic | |
| Glass bottle | |
| MFCD00005510 | |
| 21, 380 | |
| 15, 1448 | |
| Solubility in water: 14.8g/L (20°C). Other solubilities: 3.10g/100g glacial acetic acid(25°C),g/100g solvent (25°C):,tert. butanol:0.73- aceton:14.40,carbone tetrachloride:0.02- hexane:0.006,benzene:0.5 | |
| C1CC(=O)N(C1=O)Br | |
| 177.99 | |
| CHEBI:53174 | |
| 99% | |
| N-Bromosuccinimide |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
In case of major fire and large quantities: Evacuate area.
Fight fire rem
GHS Signal Word: Danger
EINECSNumber : 204-877-2
TSCA : TSCA
RUO – Research Use Only