Learn More
n-Butyl acetate, 99+%, for spectroscopy
CAS: 123-86-4 | C6H12O2 | 116.16 g/mol
$198.67 - $291.02
Chemical Identifiers
| CAS | 123-86-4 |
|---|---|
| Molecular Formula | C6H12O2 |
| Molecular Weight (g/mol) | 116.16 |
| MDL Number | MFCD00009445 |
| InChI Key | DKPFZGUDAPQIHT-UHFFFAOYSA-N |
| Synonym | n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle |
| PubChem CID | 31272 |
| ChEBI | CHEBI:31328 |
| IUPAC Name | butyl acetate |
| SMILES | CCCCOC(C)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC167685000
|
Thermo Scientific Chemicals
167685000 |
500 mL | Glass bottle |
Each for $198.67
Case of 6 Each for $0.00
|
|
||||
|
AC167680010
|
Thermo Scientific Chemicals
167680010 |
1 L | Glass bottle |
Each for $291.02
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopyChemical Identifiers
| 123-86-4 | |
| 116.16 | |
| DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
| 31272 | |
| butyl acetate |
| C6H12O2 | |
| MFCD00009445 | |
| n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
| CHEBI:31328 | |
| CCCCOC(C)=O |
Specifications
| 123-86-4 | |
| 99+% | |
| C6H12O2 | |
| MFCD00009445 | |
| 02, 130 | |
| 15, 1537 | |
| DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
| butyl acetate | |
| 31272 | |
| 116.16 | |
| ≥99.0% | |
| Liquid |
| (1 cm cell vs water), 0.01 max. at 320nm, 0.02 max. at 300nm, 0.04 max. at 275nm, 0.15 max. at 260nm, 0.4 max. at 256nm, 1 max. at 254nm | |
| Glass bottle | |
| CH3CO2(CH2)3CH3 | |
| 500 mL | |
| 0.0005% max. | |
| n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
| CCCCOC(C)=O | |
| 116.16 | |
| CHEBI:31328 | |
| 13mbar at 20°C | |
| Spectroscopy | |
| n-Butyl acetate, 99+% |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 204-658-1
RTECSNumber : AF7350000
TSCA : TSCA
RUO – Research Use Only