Learn More


n-Butyl Acetate (Reagent), Fisher Chemical™
Supplier: Thermo Fisher Scientific BPB3964
Specifications
n-Butyl Acetate | |
5°C max. | |
Colorless | |
6.2 | |
22°C | |
C6H12O2 | |
MFCD00009445 | |
1123 | |
DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
butyl acetate | |
31272 | |
116.16 | |
Reagent |
123-86-4 | |
-77.9°C | |
0.880g/cm³ | |
125°C | |
Glass Bottle | |
CH3COOCH2(CH2)2CH3 | |
4 L | |
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
CCCCOC(C)=O | |
116.16 | |
CHEBI:31328 | |
13mbar at 20°C | |
Liquid |
Chemical Identifiers
123-86-4 | |
116.16 | |
DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
31272 | |
butyl acetate |
C6H12O2 | |
MFCD00009445 | |
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
CHEBI:31328 | |
CCCCOC(C)=O |
Safety and Handling


WARNING!
Emergency Overview
Flammable liquid and vapor. May cause pulmonary edema. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.
NFPA
Health:2
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid