Learn More
n-Butyl Chloride (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations
Supplier: Fisher Chemical FLB429SK4
Description
- HPLC mobile phase for analytical and preparative separations.
- High UV transmittance and low evaporation residue.
- Filtered through a sub-micron filter to remove particulates
Specifications
n-Butyl Chloride | |
0.01 AU max., 0.01 AU max. at 254nm, 0.07 AU max. at 235nm, 0.33 AU max. at 225nm, 1.00 AU max. at 220nm | |
Colorless | |
77°C | |
Mobile phase for HPLC applications | |
Safe-Cote™ Amber Glass | |
1.3985 to 1.4015 (at 25°C) | |
MFCD00001009 | |
1127 | |
butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane | |
CCCCCl | |
92.57 | |
92.57 | |
≥99% | |
Liquid |
109-69-3 | |
-123°C | |
0.880g/cm³ | |
â12°C | |
Assay: 99% min by GC | |
C4H9Cl | |
CH3(CH2)3Cl | |
4 L | |
1ppm max. | |
VFWCMGCRMGJXDK-UHFFFAOYSA-N | |
1-chlorobutane | |
8005 | |
108mbar at 20°C | |
HPLC |
Chemical Identifiers
109-69-3 | |
92.57 | |
butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane | |
1-chlorobutane |
C4H9Cl | |
VFWCMGCRMGJXDK-UHFFFAOYSA-N | |
8005 | |
CCCCCl |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. The toxicological properties have not been fully investigated. Wear personal protective equipment. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools. Use explosion-proof equipment. To avoid ignition of vapors by static electricity discharge, all metal parts of the equipment must b. Take precautionary measures against static discharges. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.
NFPA
Health:1
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid