missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Butyl isocyanide, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 158380010

 View more versions of this product

Catalog No. AC158380010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Butyl isocyanide
2769-64-4
100.0
0.7900g/mL
98+%
C5H9N
CH3(CH2)3NC
1 g
0.79
FSBLVBBRXSCOKU-UHFFFAOYSA-N
1-isocyanobutane
76008
98+%
98+%
98.0
Colorless to Yellow
Authentic
Glass bottle
1.3950 to 1.397
MFCD00000008
11,99; 14,63
n-butyl isocyanide, butyl isocyanide, 1-butylisocyanide, n-butylisocyanide, bu-tylisonitrile, n-butylisonitrile, n-butyl isonitrile, butane,1-isocyano
CCCC[N+]#[C-]
83.13
83.13
Liquid

Chemical Identifiers

2769-64-4
83.13
FSBLVBBRXSCOKU-UHFFFAOYSA-N
76008
CCCC[N+]#[C-]
C5H9N
MFCD00000008
n-butyl isocyanide, butyl isocyanide, 1-butylisocyanide, n-butylisocyanide, bu-tylisonitrile, n-butylisonitrile, n-butyl isonitrile, butane,1-isocyano
1-isocyanobutane
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 220-458-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only