missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Butyl propionate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 269480010

 View more versions of this product

Catalog No. AC269480010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Butyl propionate
99.0
-75°C
0.8800g/mL
38°C
99+%
C7H14O2
CH3(CH2)3OC(O)C2H5
1 L
0.88
butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate
BTMVHUNTONAYDX-UHFFFAOYSA-N
butyl propanoate
11529
130.19
Liquid
590-01-2
100.0
Colorless
145°C
Authentic
Glass bottle
1.4000 to 1.402
MFCD00009448
02, 241
15, 1590
Solubility in water: 2g/L (20°C). Other solubilities: soluble in alcohol and ether
CCCCOC(=O)CC
130.19
0.83 mPa.s (20°C)
99+%

Chemical Identifiers

590-01-2
130.19
BTMVHUNTONAYDX-UHFFFAOYSA-N
11529
CCCCOC(=O)CC
C7H14O2
MFCD00009448
butyl propionate, n-butyl propionate, propanoic acid, butyl ester, n-butyl propanoate, propionic acid, butyl ester, propionic acid butyl ester, unii-2nxc4ak99e, butyl propionate natural, fema no. 2211, n-butyl n-propionate
butyl propanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-669-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only