missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Butylbenzene, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 107850500

 View more versions of this product

Catalog No. AC107850500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Butylbenzene
104-51-8
100.0
Colorless
183°C
Authentic
Glass bottle
1.4880 to 1.49
MFCD00009463
05,413
15,1551
Solubility in water: insoluble in water. Other solubilities: miscible with ethanol,ether and benzene
CCCCC1=CC=CC=C1
134.22
CHEBI:44194
99+%
99+%
99.0
-88°C
0.8600g/mL
59°C
99+%
C10H14
C6H5(CH2)3CH3
50 mL
0.86
n-butylbenzene, 1-phenylbutane, benzene, butyl, 1-butylbenzene, butyl-benzene, butane, phenyl, unii-s8xz2901rz, butylbenzene, analytical standard, n-butyl benzene, n4b
OCKPCBLVNKHBMX-UHFFFAOYSA-N
butylbenzene
7705
134.22
Liquid

Chemical Identifiers

104-51-8
134.22
OCKPCBLVNKHBMX-UHFFFAOYSA-N
7705
butylbenzene
C10H14
MFCD00009463
n-butylbenzene, 1-phenylbutane, benzene, butyl, 1-butylbenzene, butyl-benzene, butane, phenyl, unii-s8xz2901rz, butylbenzene, analytical standard, n-butyl benzene, n4b
CHEBI:44194
CCCCC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-209-7

RTECSNumber : CY9070000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only