Learn More
n-Butyllithium, 2.5M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 213351000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
n-Butyllithium | |
109-72-8 | |
0.6900g/mL | |
−21°C | |
C4H9Li | |
MFCD00009414 | |
01,95; 02,51; 04,60; 05,78; 06,85; 07,45; 08,65; 09,83; 10,68; 11,101; 12,96; 13,56; 14,63; 15,59; 16,55; 17,59 | |
n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc | |
[Li]CCCC | |
64.06 | |
64.06 |
2.5M Solution in Hexanes | |
-95.0°C | |
60.0°C to 80.0°C | |
AcroSeal™ Glass bottle | |
CH3(CH2)3Li | |
100 mL | |
0.69 | |
MZRVEZGGRBJDDB-UHFFFAOYSA-N | |
lithium;butane | |
61028 |
Chemical Identifiers
109-72-8 | |
64.06 | |
MZRVEZGGRBJDDB-UHFFFAOYSA-N | |
61028 |
C4H9Li | |
MFCD00009414 | |
n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc | |
[Li]CCCC |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. May cause drowsiness or dizziness. Suspected of damaging fertility. Highly flammable liquid and vapor. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not allow contact with air. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Store in a dry place. Store in a closed container. Ground/Bond container and receiving equipment. Take precautionary measures against static discharge.
Danger
EINECSNumber : 203-698-7
TSCA : TSCA
RUO – Research Use Only