Learn More
n-Butylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H58121AD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilityReacts with water.
Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with oxidizing agents, acid chlorides. Protect from humidity and water. Do not together with acids.
Specifications
n-Butylzinc bromide | |
92273-73-9 | |
C4H9BrZn | |
MFCD00671997 | |
UN3399 | |
bromo butyl zinc, n-butylzinc bromide, buznbr, butyl zinc bromide, n-buznbr, n-butyl zinc bromide, butylzinc bromide solution, hmbgxqklghidmn-uhfffaoysa-m, butylzinc bromide, 0.50 m in thf, butylzinc bromide solution, 0.5 m in thf | |
PRWJKDARXFAPOY-UHFFFAOYSA-M | |
bromozinc(1+);butane | |
5225082 |
Packaged under Argon in resealable ChemSeal™ bottles | |
0.958 | |
CH3(CH2)3ZnBr | |
50 mL | |
Air sensitive | |
Reacts with water. | |
CCC[CH2-].[Zn+]Br | |
202.4 | |
202.4 |
Chemical Identifiers
92273-73-9 | |
202.4 | |
PRWJKDARXFAPOY-UHFFFAOYSA-M | |
5225082 | |
CCC[CH2-].[Zn+]Br |
C4H9BrZn | |
MFCD00671997 | |
bromo butyl zinc, n-butylzinc bromide, buznbr, butyl zinc bromide, n-buznbr, n-butyl zinc bromide, butylzinc bromide solution, hmbgxqklghidmn-uhfffaoysa-m, butylzinc bromide, 0.50 m in thf, butylzinc bromide solution, 0.5 m in thf | |
bromozinc(1+);butane |
Safety and Handling
GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H225-H260-H302-H314-H335-H336-H351
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only