missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Butyric Anhydride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 108121000

 View more versions of this product

Catalog No. AC108121000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Butyric anhydride
97.5
-73.0°C
0.9630g/mL
82°C
97.5% min. (GC)
C8H14O3
(CH3CH2CH2CO)2O
100 mL
05,88
15, 1598
Solubility in water: decomposes. Other solubilities: soluble in ether,decomposes in alcohol
CCCC(=O)OC(=O)CCC
158.20
158.2
Liquid
106-31-0
100.0
Colorless to Yellow
195.0°C
Authentic
Glass bottle
1.4120 to 1.4140
MFCD00009389
02, 274
0.963
butyric anhydride, n-butyric anhydride, butanoic anhydride, butanoic acid, anhydride, butyryl oxide, butyric acid anhydride, n-butyric acid anhydride, butyranhydrid, butanoic acid anhydride, butyric anhydride n
YHASWHZGWUONAO-UHFFFAOYSA-N
butanoyl butanoate
7798
98%

Chemical Identifiers

106-31-0
158.20
YHASWHZGWUONAO-UHFFFAOYSA-N
7798
CCCC(=O)OC(=O)CCC
C8H14O3
MFCD00009389
butyric anhydride, n-butyric anhydride, butanoic anhydride, butanoic acid, anhydride, butyryl oxide, butyric acid anhydride, n-butyric acid anhydride, butyranhydrid, butanoic acid anhydride, butyric anhydride n
butanoyl butanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-383-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only