missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Carbamoyl-DL-aspartic acid, 98%, Thermo Scientific Chemicals

$692.12 - $2753.88

Chemical Identifiers

CAS 923-37-5
Molecular Formula C5H8N2O5
Molecular Weight (g/mol) 176.128
MDL Number MFCD00042822
InChI Key HLKXYZVTANABHZ-UHFFFAOYSA-N
Synonym ureidosuccinic acid, 2-ureidosuccinic acid, n-carbamoyl-dl-aspartic acid, n-aminocarbonyl-dl-aspartic acid, dl-aspartic acid, n-aminocarbonyl, 2-carbamoylamino butanedioic acid, n-carbamoylaspartic acid, carbamoyl-dl-aspartic acid, carbamylaspartic acid, 2-ureidobutanedioic acid
PubChem CID 279
ChEBI CHEBI:64850
IUPAC Name 2-(carbamoylamino)butanedioic acid
SMILES C(C(C(=O)O)NC(=O)N)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1716609
View Documents
Thermo Scientific Chemicals
A1716609
10 g
Each for $692.12
 
AAA1716618
View Documents
Thermo Scientific Chemicals
A1716618
50 g
Each for $2,753.88
 
Description

Description

N-Carbamoyl-DL-aspartic acid, is a useful biochemical for proteomics research. It is also used as a carbamate derivative which serves as an intermediate in pyrimidine biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Carbamoyl-DL-aspartic acid, is a useful biochemical for proteomics research. It is also used as a carbamate derivative which serves as an intermediate in pyrimidine biosynthesis.

Solubility
Soluble in hot water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

923-37-5
176.128
HLKXYZVTANABHZ-UHFFFAOYSA-N
279
2-(carbamoylamino)butanedioic acid
C5H8N2O5
MFCD00042822
ureidosuccinic acid, 2-ureidosuccinic acid, n-carbamoyl-dl-aspartic acid, n-aminocarbonyl-dl-aspartic acid, dl-aspartic acid, n-aminocarbonyl, 2-carbamoylamino butanedioic acid, n-carbamoylaspartic acid, carbamoyl-dl-aspartic acid, carbamylaspartic acid, 2-ureidobutanedioic acid
CHEBI:64850
C(C(C(=O)O)NC(=O)N)C(=O)O

Specifications

∼175°C (decomposition)
C5H8N2O5
1726861
Soluble in hot water.
C(C(C(=O)O)NC(=O)N)C(=O)O
10 g
279
176.13
N-Carbamoyl-DL-aspartic acid
923-37-5
MFCD00042822
ureidosuccinic acid, 2-ureidosuccinic acid, n-carbamoyl-dl-aspartic acid, n-aminocarbonyl-dl-aspartic acid, dl-aspartic acid, n-aminocarbonyl, 2-carbamoylamino butanedioic acid, n-carbamoylaspartic acid, carbamoyl-dl-aspartic acid, carbamylaspartic acid, 2-ureidobutanedioic acid
HLKXYZVTANABHZ-UHFFFAOYSA-N
2-(carbamoylamino)butanedioic acid
176.128
CHEBI:64850
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 213-096-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only