missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Carbobenzoxy-2-nitrobenzenesulfonamide 98.0+%, TCI America™
Supplier: TCI America C175725G
Specifications
N-Carbobenzoxy-2-nitrobenzenesulfonamide | |
145°C | |
C14H12N2O6S | |
25 g | |
BFQXFHUTPNBFSR-UHFFFAOYSA-N | |
benzyl N-(2-nitrophenyl)sulfonylcarbamate | |
10980593 | |
≥98.0% (HPLC,T) |
245365-64-4 | |
White | |
MFCD06797080 | |
N-Cbz-2-nitrobenzenesulfonamide | |
C1=CC=C(C=C1)COC(=O)NS(=O)(=O)C2=CC=CC=C2[N+](=O)[O-] | |
336.318 | |
336.32 | |
Crystalline Powder |
Chemical Identifiers
245365-64-4 | |
336.318 | |
BFQXFHUTPNBFSR-UHFFFAOYSA-N | |
10980593 | |
C1=CC=C(C=C1)COC(=O)NS(=O)(=O)C2=CC=CC=C2[N+](=O)[O-] |
C14H12N2O6S | |
MFCD06797080 | |
N-Cbz-2-nitrobenzenesulfonamide | |
benzyl N-(2-nitrophenyl)sulfonylcarbamate |
Safety and Handling
TSCA : No