missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Decane, 99%, Thermo Scientific Chemicals

$85.75 - $867.36

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00008954
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N
Synonym n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
PubChem CID 15600
ChEBI CHEBI:41808
IUPAC Name decane
SMILES CCCCCCCCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA14732AE
View Documents
Thermo Scientific Chemicals
A14732AE
100 mL
Each for $85.75
 
AAA14732AP
View Documents
Thermo Scientific Chemicals
A14732AP
500 mL
Each for $211.62
 
AAA147320F
View Documents
Thermo Scientific Chemicals
A147320F
2500 mL
Each for $867.36
 
Description

Description

n-Decane is used as a solvent as well as an internal standard in the GC analysis of oxalic, malonic and succinic acids. It is used in paint manufacture as a hydrocarbon solvent and for wood stains and varnishes as a varnish solvent. It is an active component of automotive fuels viz. gasoline (petrol).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Decane is used as a solvent as well as an internal standard in the GC analysis of oxalic, malonic and succinic acids. It is used in paint manufacture as a hydrocarbon solvent and for wood stains and varnishes as a varnish solvent. It is an active component of automotive fuels viz. gasoline (petrol).

Solubility
Soluble in alcohol and ethers. Miscible with ethanol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

124-18-5
142.286
DIOQZVSQGTUSAI-UHFFFAOYSA-N
15600
decane
C10H22
MFCD00008954
n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
CHEBI:41808
CCCCCCCCCC

Specifications

124-18-5
0.730
46°C (115°F)
C10H22
MFCD00008954
1696981
Soluble in alcohol and ethers. Miscible with ethanol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.
CCCCCCCCCC
142.286
CHEBI:41808
Liquid
°C to 30°C
172°C to 174°C
99%
1.4115
100 mL
n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
DIOQZVSQGTUSAI-UHFFFAOYSA-N
decane
15600
142.29
n-Decane
Videos
Safety and Handling

Safety and Handling

P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c

H226-H304

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 204-686-4

RTECSNumber : HD6550000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only