missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Dodecyl-β-D-maltoside, 99%, high purity, Thermo Scientific Chemicals
$359.74 - $4101.02
Chemical Identifiers
CAS | 69227-93-6 |
---|---|
Molecular Formula | C24H46O11 |
Molecular Weight (g/mol) | 510.6 |
MDL Number | MFCD00043012 |
InChI Key | NLEBIOOXCVAHBD-QKMCSOCLSA-N |
Synonym | n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside |
PubChem CID | 114880 |
ChEBI | CHEBI:43769 |
IUPAC Name | (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
SMILES | CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC329370010
|
Thermo Scientific Chemicals
329370010 |
1 g | Glass bottle |
Each for $359.74
|
|
|||||
AC329370050
|
Thermo Scientific Chemicals
329370050 |
5 g | Glass bottle |
Each for $1,192.43
|
|
|||||
AC329370250
|
Thermo Scientific Chemicals
329370250 |
25 g | Glass bottle |
Each for $4,101.02
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69227-93-6 | |
510.6 | |
NLEBIOOXCVAHBD-QKMCSOCLSA-N | |
114880 | |
(2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
C24H46O11 | |
MFCD00043012 | |
n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside | |
CHEBI:43769 | |
CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O |
Specifications
69227-93-6 | |
100.0 | |
White | |
98.5% min. (ELSD) (HPLC) | |
C24H46O11 | |
1 g | |
+47.50 (20.00°C c=1,water) | |
n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside | |
NLEBIOOXCVAHBD-QKMCSOCLSA-N | |
(2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
114880 | |
510.6 | |
High Purity | |
n-Dodecyl-β-D-maltoside |
98.5 | |
217.0°C | |
Authentic | |
Glass bottle | |
MFCD00043012 | |
17, V,8, 12 | |
+ 47.50 | |
Solubility in water: soluble (100g/L). Other solubilities: soluble in methanol, ethanol | |
CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O | |
510.6 | |
CHEBI:43769 | |
99% | |
Powder |
RUO – Research Use Only