Learn More
N-Dodecylmethylamine, 98%
CAS: 7311-30-0 | C13H29N | 199.382 g/mol
$74.32 - $300.10
Chemical Identifiers
| CAS | 7311-30-0 |
|---|---|
| Molecular Formula | C13H29N |
| Molecular Weight (g/mol) | 199.382 |
| MDL Number | MFCD00026574 |
| InChI Key | OMEMQVZNTDHENJ-UHFFFAOYSA-N |
| Synonym | n-methyldodecylamine, dodecyl methyl amine, dodecylmethylamine, 1-dodecanamine, n-methyl, n-dodecylmethylamine, dodecyl-methyl-amine, unii-88nn43i94s, methyldodecylamine, n-methyldodecanamine, n-methyl-n-dodecylamine |
| PubChem CID | 81746 |
| IUPAC Name | N-methyldodecan-1-amine |
| SMILES | CCCCCCCCCCCCNC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0702503
|
Thermo Scientific Chemicals
L0702503 |
1 g |
Each for $74.32
|
|
|||||
|
AAL0702506
|
Thermo Scientific Chemicals
L0702506 |
5 g |
Each for $300.10
|
|
|||||
Description
N-Dodecylmethylamine is used as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Dodecylmethylamine is used as intermediates.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
Chemical Identifiers
| 7311-30-0 | |
| 199.382 | |
| OMEMQVZNTDHENJ-UHFFFAOYSA-N | |
| 81746 | |
| CCCCCCCCCCCCNC |
| C13H29N | |
| MFCD00026574 | |
| n-methyldodecylamine, dodecyl methyl amine, dodecylmethylamine, 1-dodecanamine, n-methyl, n-dodecylmethylamine, dodecyl-methyl-amine, unii-88nn43i94s, methyldodecylamine, n-methyldodecanamine, n-methyl-n-dodecylamine | |
| N-methyldodecan-1-amine |
Specifications
| 7311-30-0 | |
| 90°C to 92°C (0.9 mmHg) | |
| C13H29N | |
| MFCD00026574 | |
| UN2735 | |
| n-methyldodecylamine, dodecyl methyl amine, dodecylmethylamine, 1-dodecanamine, n-methyl, n-dodecylmethylamine, dodecyl-methyl-amine, unii-88nn43i94s, methyldodecylamine, n-methyldodecanamine, n-methyl-n-dodecylamine | |
| OMEMQVZNTDHENJ-UHFFFAOYSA-N | |
| N-methyldodecan-1-amine | |
| 81746 | |
| 98% |
| 0.795 | |
| >110°C (230°F) | |
| 1.4395 | |
| 1 g | |
| 1739959 | |
| Not miscible in water. | |
| CCCCCCCCCCCCNC | |
| 199.382 | |
| 199.38 | |
| N-Dodecylmethylamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 230-771-0
TSCA : No
RUO – Research Use Only