missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Ethyl-1-naphthylamine Hydrobromide 98.0+%, TCI America™
Supplier: TCI America E014925G
Specifications
N-Ethyl-1-naphthylamine Hydrobromide | |
White-Yellow | |
MFCD00060154 | |
SBGSNWFFTCHTQG-UHFFFAOYSA-N | |
N-ethylnaphthalen-1-aminium bromide | |
44148659 | |
≥98.0% (T) |
36966-04-8 | |
C12H14BrN | |
25 g | |
[Br-].CC[NH2+]C1=C2C=CC=CC2=CC=C1 | |
252.16 | |
252.16 | |
Crystalline Powder |
Chemical Identifiers
36966-04-8 | |
252.16 | |
SBGSNWFFTCHTQG-UHFFFAOYSA-N | |
N-ethylnaphthalen-1-aminium bromide |
C12H14BrN | |
MFCD00060154 | |
44148659 | |
[Br-].CC[NH2+]C1=C2C=CC=CC2=CC=C1 |
Safety and Handling
EINECSNumber : (4)-0349
TSCA : No