missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Ethyldiisopropylamine, 99%, Thermo Scientific Chemicals

$87.19 - $927.89

Chemical Identifiers

CAS 7087-68-5
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008868
InChI Key JGFZNNIVVJXRND-UHFFFAOYSA-N
Synonym n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
PubChem CID 81531
IUPAC Name N-ethyl-N-propan-2-ylpropan-2-amine
SMILES CCN(C(C)C)C(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA11801AE
View Documents
Thermo Scientific Chemicals
A11801AE
100 mL
Each for $87.19
 
AAA11801AP
View Documents
Thermo Scientific Chemicals
A11801AP
500 mL
Each for $284.40
 
AAA118010F
View Documents
Thermo Scientific Chemicals
A118010F
2500 mL
Each for $927.89
 
Description

Description

N-Ethyldiisopropylamine is used to make or formulate industrial products. It is also used in organic chemistry as a base. It acts as a selective reagent in the alkylation of secondary amines to tertiary amines by alkyl halides. It reacts with monoactivated Michael acceptors to afford symmetrical sulfones. Further, it is used to prepare heterocyclic compound, sscorpionine by a reaction with disulfur dichloride using 1,4-diazabicyclo[2.2.2]octane (DABCO) as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethyldiisopropylamine is used to make or formulate industrial products. It is also used in organic chemistry as a base. It acts as a selective reagent in the alkylation of secondary amines to tertiary amines by alkyl halides. It reacts with monoactivated Michael acceptors to afford symmetrical sulfones. Further, it is used to prepare heterocyclic compound, sscorpionine by a reaction with disulfur dichloride using 1,4-diazabicyclo[2.2.2]octane (DABCO) as a catalyst.

Solubility
Miscible with water and ethanol.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, carbon dioxide, copper, brass, rubber, oxidizing agents, nitrates, peroxides, aldehydes and metals.
Specifications

Chemical Identifiers

7087-68-5
129.247
JGFZNNIVVJXRND-UHFFFAOYSA-N
81531
CCN(C(C)C)C(C)C
C8H19N
MFCD00008868
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
N-ethyl-N-propan-2-ylpropan-2-amine

Specifications

7087-68-5
0.747
8°C (46°F)
C8H19N
MFCD00008868
605301
Miscible with water and ethanol.
CCN(C(C)C)C(C)C
129.247
129.25
N-Ethyldiisopropylamine
−46°C
126°C to 127°C
99%
1.4135
100 mL
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
JGFZNNIVVJXRND-UHFFFAOYSA-N
N-ethyl-N-propan-2-ylpropan-2-amine
81531
Liquid
Videos
Safety and Handling

Safety and Handling

P210-P233-P234-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P330-P370+P378q-P390-P501c

H225-H290-H302-H318-H331-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: I

EINECSNumber : 230-392-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only